Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MPEP hydrochloride

Potent mGlu5 antagonist; also positive allosteric modulator of mGlu4 receptors

$220.40 - $890.88

Specifications

Chemical Name or Material 2-Methyl-6-(phenylethynyl)pyridine hydrochloride
CAS 219911-35-0
Assay Percent Range >99%
Solubility Soluble to 100mM in DMSO
Target Group I mGluR Antagonists
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
12-121-0
View Documents
Tocris Bioscience™
1212/10
10 mg
Each for $220.40
Add to Cart
 
12-125-0R
View Documents
Tocris Bioscience™
1212/50
50 mg
Each for $890.88
Add to Cart
 
Description

Description

Potent and highly selective non-competitive antagonist at the mGlu5 receptor subtype (IC50 = 36 nM) and a positive allosteric modulator at mGlu4 receptors. Centrally active following systemic administration in vivo. Reverses mechanical hyperalgesia in the inflamed rat hind paw.
Specifications

Specifications

2-Methyl-6-(phenylethynyl)pyridine hydrochloride
>99%
Group I mGluR Antagonists
Observed MW: 229.71
219911-35-0
Soluble to 100mM in DMSO
C14H11N.HCl
99%
Safety and Handling

Safety and Handling

Recommended Storage : Desiccate at +4°C

SDS
Documents

Documents

Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit