Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mPlum Goat anti-Tag, Polyclonal, liquid, TrueMAB™

Goat Polyclonal Antibody
Supplier: OriGene TA150098

Description
mPlum fluorescent protein.Specifications
mPlum | |
Polyclonal | |
Whole serum with 0.05% sodium azide | |
Goat | |
150 μL | |
Primary | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
mPlum | |
Purified mPlum protein expressed in E.coli | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction