Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPND Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19153620UL

 View more versions of this product

Catalog No. NBP19153620


Add to cart

Description

Description

MPND Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

MPND
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
MPND
Synthetic peptide directed towards the middle region of human E130307M08RIK. Peptide sequence EGEEEELLLEEEEEDVLAGVSSEDKGHRPPGKGSLEPEATPPGKRMDKVP.
48 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Western Blot
Western Blot 1:1000
NP_080806
EC 3.4, FLJ14981, MPN domain containing, MPN domain-containing protein
Rabbit
IgG
Protein A purified
RUO
Primary
84954
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit