Learn More
CAPRICO BIOTECHNOLOGIES INC MPO PE 4123025
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4123025
Encompass_Preferred
The MPO-PE 4123025 is a polyethylene foam sheet from Caprico Bio. It is a closed cell, crosslinked polyethylene foam with a density of 2.2 lbs/ft3. The sheet has a thickness of 1/4" and comes in a 48" x 96" size. It has excellent water resistance and low thermal conductivity. The foam provides good energy absorption and has high tensile strength. It can withstand temperatures from -70 to 200°F. The polyethylene foam sheet has applications including packaging, cushioning, insulation, floatation, and more.
Catalog No. 50-227-9320
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.