Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
BioVision IncSupplier Diversity Partner Myeloperoxidase (MPO) Peroxidation Activity Assay Kit: High-Throughput suitable Assay for Measuring MPO Activity in Tissues, Cells etc. Simple, Sensitive and Reliable. Detection Limit: <2 μU. 100 Assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K747100
Myeloperoxidase (MPO) Peroxidation Activity Assay Kit: Myeloperoxidase (MPO: EC 1.11.2.2) catalyzes the hydrogen peroxide (H2O2) dependent oxidation of chloride anions (Cl-) to generate hypochlorite (HClO). During respiratory burst, neutrophils utilize MPO activity to exhibit cytotoxicity against tumor cells and pathogenic microorganisms. BioVision's Myeloperoxidase (MPO) Peroxidation Activity Assay Kit, oxidizes a substrate to generate fluorescence (Ex/Em = 535/587 nm), which is directly proportional to total peroxidase activity in the sample. A specific MPO inhibitor is provided, which suppresses peroxidase activity due to MPO. This permits differentiation of MPO-mediated peroxidation from the activity of other peroxidases which may be present in the samples. Our kit provides a sensitive, quick and easy method for detecting MPO activity. The assay is high-throughput adaptable and can detect less than 2 ¿U of MPO activity. BioVision charges an Ice pack fee for each order.
Catalog No. 50-111-9875