Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPP7 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549335

Catalog No. PA549335

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Acts as an important adapter that promotes epithelial cell polarity and tight junction formation via its interaction with DLG1. Involved in the assembly of protein complexes at sites of cell-cell contact.
Specifications

Specifications

MPP7
Polyclonal
PBS with 0.09% sodium azide
Q5T2T1, Q8BVD5
MAGUK p55 subfamily member 7, MPP7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
143098, 75739
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MPP7
Liquid
MPP7
KLH conjugated synthetic peptide between 2-36 amino acids from the N-terminal region of human MPP7.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit