Learn More
MPP8 Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303052AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Heterochromatin component that specifically recognizes and binds methylated ′Lys-9′ of histone H3 (H3K9me) and promotes recruitment of proteins that mediate epigenetic repression (PubMed:20871592, PubMed:26022416). Mediates recruitment of the HUSH complex to H3K9me3 sites: the HUSH complex is recruited to genomic loci rich in H3K9me3 and is probably required to maintain transcriptional silencing by promoting recruitment of SETDB1, a histone methyltransferase that mediates further deposition of H3K9me3 (PubMed:26022416). Binds H3K9me and promotes DNA methylation by recruiting DNMT3A to target CpG sites; these can be situated within the coding region of the gene (PubMed:20871592). Mediates down-regulation of CDH1 expression (PubMed:20871592).Specifications
MPP8 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q99549 | |
MPHOSPH8 | |
Between 200 and 250 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MPHOSPH8 | |
HSMPP8, TWA3, mpp8, M-phase phosphoprotein, mpp, M-phase phosphoprotein, mpp8, two hybrid-associated protein 3 with RanBPM | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54737 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.