Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPPED2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549036

Catalog No. PA549036

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with rat based on sequence homology.

Displays low metallophosphoesterase activity (in vitro). May play a role in the development of the nervous system.
Specifications

Specifications

MPPED2
Polyclonal
PBS with 0.09% sodium azide
Q15777
Metallophosphoesterase MPPED2, 31--, Fetal brain protein 239, 239FB, Metallophosphoesterase domain-containing protein 2, MPPED2, C11orf8, FAM1B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
744
Western Blot
Unconjugated
MPPED2
Liquid
MPPED2
KLH conjugated synthetic peptide between 224-251 amino acids from the C-terminal region of human MPPED2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit