Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPPG, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
169209-65-8
C9H12NO5P
245.171
PAONCRJPUQXPRW-UHFFFAOYSA-N
mppg,2-amino-2-4-phosphonophenyl propanoic acid,chembl86508,d0d2gl,mppg nmr , solid,rs-?-methyl-4-phosphonophenylglycine,rs-,a-methyl-4-phosphonophenylglycine,2-amino-2-4-phosphono-phenyl-propionic acid,2-amino-2-4-phosphono-phenyl-propionic acid rs-mppg
3972752
2-amino-2-(4-phosphonophenyl)propanoic acid
CC(C1=CC=C(C=C1)P(=O)(O)O)(C(=O)O)N
169209-65-8
C9H12NO5P
245.171
PAONCRJPUQXPRW-UHFFFAOYSA-N
mppg,2-amino-2-4-phosphonophenyl propanoic acid,chembl86508,d0d2gl,mppg nmr , solid,rs-?-methyl-4-phosphonophenylglycine,rs-,a-methyl-4-phosphonophenylglycine,2-amino-2-4-phosphono-phenyl-propionic acid,2-amino-2-4-phosphono-phenyl-propionic acid rs-mppg
3972752
2-amino-2-(4-phosphonophenyl)propanoic acid
CC(C1=CC=C(C=C1)P(=O)(O)O)(C(=O)O)N

Group III/group II mGlu antagonist. More selective for group III than group II

Potent antagonist of L-AP4-induced effects in rat spinal cord, thalamic and hippocampal neurons, showing selectivity over (1S,3S)-ACPD-induced effects. Potent antagonist of mGlu receptors linked to adenylyl cyclase in adult rat cortical slices.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Formula Weight 245.17
Percent Purity >98%
Quantity 10mg
Chemical Name or Material MPPG

Recommended Storage : Store at Room Temperature