Learn More
Tocris Bioscience™ Mps BAY 2a
Potent and selective Mps1 kinase inhibitor
$239.00 - $1009.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
556250 View Documents Tocris Bioscience™
5562/50
Each for $1,009.00
Add to cart
556210 View Documents Tocris Bioscience™
5562/10
Each for $239.00
Add to cart
Description
Description
Potent and selective Mps1 inhibitor (IC50 = 1 nM for human enzyme). Selective for Mps1 over a panel of 220 kinases. Differentially inhibits human colon carcinoma cell proliferation. Blocks cell cycle progression by preventing spindle assembly checkpoint activation. Induces apoptosis.