Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MPure-12™ Automated Nucleic Acid Purification System

Purifies nucleic acids from a wide variety of bio-specimens using magnetic bead separation technology

Manufacturer:  MP Biomedicals™ 117002200

Catalog No. 117002200


Add to cart

Includes: MPure-12 Instrument with Bar Code Reader

Description

Description

  • MPure-12 is a bench-top automated system
  • Purifies nucleic acids from a wide variety of bio-specimens using magnetic bead separation technology.
  • It has a uniquely designed magnetic bead processing chamber.
  • Fully integrated with easy to use pre-packaged reagent kits.

Certifications

CE

Specifications

Specifications

Made of Metal, Glass, Plastic
70cm
20.4 in.
Magnetic Bead Separation Technology
Instruments and Systems
Blood, Tissues, Cultured Cells, FFPE, Bacteria, Serum, Plasma, Swabs, Plants, Plant and Forensic samples
59 to 86°F
5 IU spiked (about 1 IU in PCR reaction)
110 to 240V
27.5 in.
Nucleic Acid Purification (DNA, RNA)
52cm
1 to 12 samples
100 to 1000μL of sample
Room Temperature
15 to 30°C
Run Time: 30 to 70 minutes
Sample Volume: 50 to 1000μL
Elution Volume: 50 to 300μL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit