Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MPure™ Cultured Cell DNA Extraction Kit

For extraction of genomic DNA from cultured cells in monolayer or suspension and from buffy coat

Manufacturer:  MP Biomedicals™ 117022500

Catalog No. 11-702-2500


Add to cart

Description

Description

  • Completely automated purification protocol
  • Run times as little as 40 minutes
  • DNA can be used immediately in downstream applications such as PCR, qPCR, Sequencing (NGS), Microarray, RFLP and Southern Blot Analysis

Specifications

Specifications

Cultured Cell DNA Extraction Kit
MPure-12 Automated Nucleic Acid Purification System
Magnetic Bead Technology
Do not use more than 5 x 106 cells with a normal set of chromosomes
Store at room temperature
DNA Purification
Proteinase K solution, 40μL; Lysis Buffer 2, 720μL; Binding Buffer 1, 720μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL
48 preps
Cultured Cells and Buffy Coat
As little as 40 minutes
Safety and Handling

Safety and Handling

ShelfLife : 12 months

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit