Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MPure™ Viral/Pathogen Nucleic Acids Extraction Kit A

Extract viral and bacterial DNA/RNA from cell-free samples including serum, plasma and other cell-free body fluids

Manufacturer:  MP Biomedicals™ 117022120

Catalog No. 11-702-2120


Add to cart

Description

Description

  • Run in as little as 40 minutes
  • Nucleic acid is ready for immediate use

Specifications

Specifications

Viral/Pathogen Nucleic Acid Extraction Kit
MPure-12 Automated Nucleic Acid Purification System
48 preps
Store at room temperature
Nucleic Acid Purification, DNA and RNA
Reagent Cartridge: 48; Reaction Chamber: 48; Tip Holder: 48; Filtered Tip: 50; Piercing Pin: 50; Sample Tube (2mL): 50; Elute Tube (1.5mL): 50; RNA Carrier (1mg): 1; Barcode Paper: 1; Proteinase K solution: 40μL; Lysis Buffer 1: 720μL; Binding Buffer 1: 1000μL; Magnetic Bead Solution: 800μL; Washing Buffer 1: 1000μL; Washing Buffer 2: 1000μL; Washing Buffer 3: 1000μL; RNase-free water: 1000μL; BL2 Buffer: 400μL
100-400μL (virus); 100-200μL (virus/bacteria)
As little as 40 minutes
Safety and Handling

Safety and Handling

ShelfLife : 12 months

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit