Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MPure™ Viral RNA Extraction Kit

Extract Viral RNA from human biological specimens such as serum, plasma, urine, CSF and other cell-free fluids

Manufacturer:  MP Biomedicals™ 117022140

Catalog No. 11-702-2140


Add to cart

Description

Description

  • Completely automated purification protocol
  • Run times as little as 40 minutes
  • RNA can be used immediately in downstream applications

Specifications

Specifications

Viral RNA Extraction Kit
MPure-12 Automated Nucleic Acid Purification System
Magnetic Bead Technology
100 to 400μL
Store at room temperature
RNA Purification
Proteinase K, 30μL; Lysis Buffer 2, 720μL; Binding Buffer 1, 1000μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL; RNase Free water, 1000μL; RNase Free water, 1000μL
48 preps
Human biological specimens such as serum, plasma, and other cell-free fluids
Safety and Handling

Safety and Handling

ShelfLife : 12 months

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit