Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MPV17L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560245
Description
Immunogen sequence: GCLEGQTVGE SCQELREKFW EFYKADWCVW PAAQFVNFLF VPPQFRVTYI NGLTLGWDTY LSYLKYRSPV PLTPPGCVAL DTRAD Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.
FKSG24 is a multi-pass membrane proteinPotential. It belongs to the peroxisomal membrane protein PXMP2/4 family. The exact function of FKSG24 remains unknown.
Specifications
MPV17L2 | |
Polyclonal | |
Unconjugated | |
MPV17L2 | |
FKSG24; MPV17 mitochondrial inner membrane protein like 2; MPV17 mitochondrial membrane protein-like 2; MPV17L2; mpv17-like protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84769 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q567V2 | |
MPV17L2 | |
Recombinant protein corresponding to Human MPV17L2. Recombinant protein control fragment (Product #RP-99025). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction