Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MPZL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
$337.75 - $627.50
Specifications
Antigen | MPZL1 |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:10-1:20 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MPZL1 Polyclonal specifically detects MPZL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MPZL1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
FLJ21047, immunoglobulin family transmembrane protein, MPZL1b, myelin protein zero-like 1, myelin protein zero-like protein 1, protein zero related, Protein zero-related, PZR1b, PZRb, PZRPZRa | |
MPZL1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:10-1:20 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
9019 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:KEIFVANGTQGKLTCKFKSTSTTGGLTSVSWSFQPEGADTTVSFFHYSQGQVYLGNYPPFKDRISWAGDLDKKDASINIENMQFIHNGTYICDVKNPPDIVVQPGHIRLYVVEKENLPVFPVWVV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title