Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rae-1 Rat anti-Mouse, Alexa Fluor 700, Clone: 186107, R&D Systems
Rat Monoclonal Antibody
Manufacturer: R&D Systems FAB17582N100UG
This item is not returnable.
View return policy
Description
Rae-1 Monoclonal antibody specifically detects Rae-1 in Mouse samples. It is validated for Flow Cytometry.Specifications
Rae-1 | |
186107 | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q9JI58 | |
Raet1a | |
Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta , Leu29-Ser227, Accession # Q9JI58 | |
Protein A or G purified from hybridoma culture supernatant | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
Monoclonal | |
Detects mouse Rae-1. It recognizes Rae-1 alpha , beta , delta , gamma and epsilon . |
Flow Cytometry | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Conjugated, Purified, Supernatant | |
Rat | |
IgG2a | |
100 μg | |
Primary | |
8480 | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title