Learn More
Heidolph™ MR Hei-Tec Magnetic Hotplate Stirrer Package with PT1000 PROMO
For smooth to intense mixing and heating of low-viscosity fluids. An excellent choice for decomposing organic and inorganic substances.
Manufacturer:  Heidolph™ 036810584
 View more versions of this product
Catalog No. 05-405-409PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
All Units Feature
  • For smooth to intense mixing and heating of low-viscosity fluids, an excellent choice for decomposing organic and inorganic substances.
  • Hotplate reaches a maximum temperature of 300°C
  • For your protection an independent safety circuit will switch off heating if hotplate temperature exceeds 25°C over set temperature
  • A separate on/off switch for heating prevents an unintentional heat-up. If heating is switched on the on/off button is illuminated for visual control
  • Strong magnetic field enables stirring even high viscosity media or volumes of 20L of water with ease
  • Speed is variably adjustable from 100 to 1400 rpm to an accuracy of ±2% for gentle mixing
Kera-Disk top plate- fast heat-up times and chemical resistance
  • Glass ceramic top plates offer the benefits of chemical-resistance, scratch-proof but produce longer heat-up times
  • Aluminum top plates allow for immediate heat transfer but are not chemical-resistant or scratch-proof
  • Kera-Disk top plates take the best of two worlds: The aluminum top plate allows of immediate heat transfer for quick heat-up times and a thin layer of ceramic coating makes the top plates chemical-resistant and scratch-proof
Sealed Housing- guarantees extended performance life and reduced maintenance
  • Protection against aggressive fumes, liquids and vapors which prevents internal corrosion
  • This results in an increased lifespan of 10 years on average at reduced maintenance and repair cost
  • Protection of all mechanical and electronic components against aggressive environmental conditions
Hei-Tec
  • For high requirements and reproducible results
  • Features a digital display and allows for easy setting and monitoring of values in the illuminated readout
  • Digital speed setting
  • Pt 1000 temperature sensor for precise temperature control, overshoot protection and reproducible results
Specifications
Specifications
Ceramic Coated Aluminum
Temp sensor; Rod; Clamp; Digital speed settings; Digital display
20°C to 300°C
100 to 1400 rpm
115 V
135 mm dia.
Hot Plate Stirrer, Stirrer
Digital