Learn More
Medchemexpress LLC MR-L2 | 2374703-19-0 | 98.63% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12835810MG
Encompass_Preferred
MR-L2 is a reversible and noncompetitive allosteric activator of long-isoform phosphodiesterase-4 (PDE4). It activates representative PDE4 long-isoform variants (PDE4A4, PDE4B1, PDE4C3, PDE4D5). This compound suppresses PGE2-induced MDCK cell cyst formation with an EC50 of 1.2 μM and significantly suppresses cAMP elevation and cysts formation in MDCK cells at concentrations of 0.3-10 μM. MR-L2 suppresses the number of cysts formation in 3D culture for both unstimulated and vasopressin-treated culture conditions, without affecting cell viability.
  • Reversible and noncompetitive allosteric activator of long-isoform phosphodiesterase-4 (PDE4)
  • Activates representative PDE4 long-isoform variants
  • Suppresses PGE2-induced MDCK cell cyst formation with an EC50 of 1.2 μM
  • Significantly suppresses cAMP elevation and cysts formation in MDCK cells
  • Suppresses cysts formation in 3D culture without affecting cell viability
Catalog No. 50-003-39301