Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRCK alpha Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575816
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
CDC42BPA (MRCKA) is a serine/threonine kinase which functions in actin cytoskeleton assembly and organization. Three independent coiled-coil domains and the N-terminal region preceding the kinase domain are responsible for intermolecular interactions leading to MRCK alpha multimerization. The transautophosphorylation process is critical for regulation of MRCK alpha catalytic activities. Binding of phorbol esters to MRCK releases its autoinhibition, allowing N-terminal dimerization and subsequent kinase activation.
Specifications
MRCK alpha | |
Polyclonal | |
Unconjugated | |
CDC42BPA | |
A930014J19Rik; CDC42 binding protein kinase alpha; CDC42 binding protein kinase alpha (DMPK-like); CDC42 binidng protein kinase beta; CDC42-binding protein kinase alpha; CDC42-binding protein kinase alpha (DMPK-like); CDC42BPA; cdc42bpa {ECO:0000250; DKFZp686L1738; DKFZp686P1738; DMPK-like; DMPK-like alpha; FLJ23347; Kiaa0451; MRCK; MRCK alpha; MRCKA; myotonic dystrophy kinase-related CDC42-binding kinase alpha; myotonic dystrophy kinase-related CDC-42-binding kinase-alpha; myotonic dystrophy kinase-related CDC42-binding protein kinase alpha; myotonic dystrophy protein kinase-like alpha; mytonic dystrophy kinase-related Cdc42-binding kinase; PK428; RP5-1087E8.4; serine/threonine-protein kinase MRCK alpha; ser-thr protein kinase PK428; ser-thr protein kinase related to the myotonic dystrophy protein kinase; UniProtKB:Q5VT25} | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
226751, 8476 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3UU96, Q5VT25 | |
CDC42BPA | |
Synthetic peptide corresponding to residues in Human MRCK alpha. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction