Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRCK Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: OTI12C12, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272777V
Description
MRCK Monoclonal specifically detects MRCK in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MRCK | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CDC42BPA | |
Recombinant protein fragment corresponding to amino acids 1419-1638 of human MRCK (NP_055641) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI12C12 | |
Western Blot | |
CDC42 binding protein kinase alpha (DMPK-like), CDC42 binidng protein kinase beta, CDC42-binding protein kinase alpha, CDC42-binding protein kinase alpha (DMPK-like), DMPK-like alpha, EC 2.7.11, EC 2.7.11.1, KIAA0451DKFZp686L1738, MRCK, MRCK alpha, MRCKADKFZp686P1738, Myotonic dystrophy kinase-related CDC42-binding kinase alpha, myotonic dystrophy kinase-related CDC42-binding protein kinase alpha, Myotonic dystrophy protein kinase-like alpha, PK428FLJ23347, serine/threonine-protein kinase MRCK alpha, ser-thr protein kinase PK428, ser-thr protein kinase related to the myotonic dystrophy protein kinase | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
8476.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title