Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mre11 Armenian Hamster anti-Human, Mouse, Allophycocyanin/Cy7, Clone: 15B8.1E7.6, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259677APCCY7

Catalog No. N259677APC7


Add to cart

Description

Description

Mre11 Monoclonal antibody specifically detects Mre11 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

Mre11
15B8.1E7.6
Flow Cytometry
Conjugated, Purified
MRE11
Paritial recombinant mouse Mre11 protein (amino acids 68-608) [UniProt Q61216]
Protein A or G purified
Breast Cancer, Cancer, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Protein Phosphatase
Primary
4361.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11MRE11B
Armenian Hamster
IgG
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit