Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mre11 Hamster anti-Human, Mouse, Clone: 15B8.1E7.6, Novus Biologicals™

Hamster MonoclonalAntibody

$99.50 - $305.50

Specifications

Antigen Mre11
Clone 15B8.1E7.6
Dilution Western Blot 1 - 2 ug/ml, Flow Cytometry 1 - 2.5 ug/mL, Immunohistochemistry 2 ug/ml, Immunocytochemistry/Immunofluorescence 5-10 ug/ml, Immunohistochemistry-Paraffin 2 ug/ml, Flow (Intracellular)
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
NBP259677 View Documents Novus BiologicalsSupplier Diversity Partner
NBP259677
0.1 mg Unconjugated Each for $305.50
Add to cart
 
NBP259677SS View Documents Novus BiologicalsSupplier Diversity Partner
NBP259677SS
0.025 mg Unconjugated Each for $99.50
Add to cart
 
Description

Description

Mre11 Monoclonal antibody specifically detects Mre11 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications

Specifications

Mre11
Western Blot 1 - 2 ug/ml, Flow Cytometry 1 - 2.5 ug/mL, Immunohistochemistry 2 ug/ml, Immunocytochemistry/Immunofluorescence 5-10 ug/ml, Immunohistochemistry-Paraffin 2 ug/ml, Flow (Intracellular)
Purified
Human, Mouse
PBS with 0.02% Sodium Azide
Paritial recombinant mouse Mre11 protein (amino acids 68-608) [UniProt Q61216]
Monoclonal
Protein A or G purified
15B8.1E7.6
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Armenian Hamster
RUO
4361
IgG
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit