Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRFAP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19855220UL
Description
MRFAP1 Polyclonal specifically detects MRFAP1 in Human samples. It is validated for Western Blot.Specifications
MRFAP1 | |
Polyclonal | |
Western Blot 1:1000 | |
NP_150638 | |
MRFAP1 | |
The immunogen for this antibody is MRFAP1 - N-terminal region. Peptide sequence PVINEMREDIASLTREHGRAYLRNRSKLWEMDNMLIQIKTQVEASEESAL. | |
Affinity Purified | |
RUO | |
93621 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Mof4 family associated protein 1, Morf4 family associated protein 1, PAM14MORF4 family-associated protein 1, PGR1protein associated with MRG, 14 kDa, Protein associated with MRG of 14 kDa, Protein PGR1, T-cell activation protein | |
Rabbit | |
15 kDa | |
20 μL | |
Primary | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title