Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mRFP Mouse anti-Tag, Magnetic beads, Clone: OTI3D7, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA183043

Description
mRFP (red fluorescent protein) fluorescent protein.Specifications
mRFP | |
Monoclonal | |
PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide | |
Mouse | |
Affinity Chromatography | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Immunoprecipitation | |
OTI3D7 | |
mRFP | |
Recombit protein of mRFP expressed in HEK293 cells. | |
1 mL | |
Primary | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction