Learn More
MRGPRF Mouse anti-Human, Alexa Fluor 350, Clone: 916219, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8396U100UG
Catalog No. FAB8396U100
Add to cart
This item is not returnable. View return policy
Description
Description
MRGPRF Monoclonal antibody specifically detects MRGPRF in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
MRGPRF
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
NP_659452
MRGPRF
NS0 mouse myeloma cell line transfected with human MRGPRF Met1-Ser343 Accession # NP_659452
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects HEK293 human embryonic kidney cell line transfected with human MRGPRF by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells.
Flow Cytometry
916219
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
FLJ16111, FLJ29034, FLJ40998, FLJ53714, G protein-coupled receptor 140, G protein-coupled receptor 168, G protein-coupled receptor MrgF, GPR140, GPR168, G-protein coupled receptor 140, G-protein coupled receptor 168, mas-related G protein-coupled MRGF, Mas-related gene F protein, MAS-related GPR, member F, mas-related G-protein coupled receptor member F, MGC21621, mrgF, MRGFDKFZp586B2122, RTA, seven transmembrane helix receptor
Mouse
IgG1
RUO
Primary
116535
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)