Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRGPRX4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521918
Description
Recommended positive controls: H1299, HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Orphan receptor. Probably involved in the function of nociceptive neurons. May regulate nociceptor function and/or development, including the sensation or modulation of pain. Potently activated by enkephalins.
Specifications
MRGPRX4 | |
Polyclonal | |
Unconjugated | |
MRGPRX4 | |
G protein-coupled receptor MRGX4; G protein-coupled receptor SNSR5; G protein-coupled receptor SNSR6; GPCR; MAS related GPR family member X4; MAS-related GPR, member X4; mas-related G-protein coupled receptor member X4; MGC129753; MGC129754; MRGPRX4; MRGX4; sensory neuron-specific G-protein coupled receptor 5/6; SNSR5; SNSR6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117196 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q96LA9 | |
MRGPRX4 | |
Synthetic peptide corresponding to a region within amino acids 258 and 322 of MRGPRX4 (Uniprot ID#Q96LA9) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction