Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRI1 Monoclonal Antibody (OTI6A10), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524932
Description
MRI1 Monoclonal antibody specifically detects MRI1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
MRI1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BV20 | |
MRI1 | |
Full length human recombinant protein of MRI1 produced in HEK293T cell | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI6A10 | |
Unconjugated | |
MRI1 | |
2410018C20Rik; CH73-131E21.1; HAMAP-Rule:MF_03119}; M1Pi; M1Pi {ECO:0000255; mediator of RhoA-dependent invasion; Methylthioribose-1-phosphate isomerase; methylthioribose-1-phosphate isomerase {ECO:0000255; methylthioribose-1-phosphate isomerase 1; methylthioribose-1-phosphate isomerase homolog; methylthioribose-1-phosphate isomerase homolog (S. cerevisiae); MGC3207; MRDI; Mri1; MTNA; MTR-1-P isomerase; MTR-1-P isomerase {ECO:0000255; RGD1307789; similar to CG11334-PB, isoform B; S-methyl-5-thioribose-1-phosphate isomerase; S-methyl-5-thioribose-1-phosphate isomerase {ECO:0000255; S-methyl-5-thioribose-1-phosphate isomerase 1; translation initiation factor eIF-2B subunit alpha/beta/delta-like protein; translation initiation factor eIF-2B subunit alpha/beta/delta-like protein {ECO:0000255; UNQ6390/PRO21135; Ypr118w; zgc:172216 | |
Mouse | |
Affinity Chromatography | |
RUO | |
84245 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction