Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRP1 Mouse anti-Human, Clone: MRP1/1343, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30733210UG

 View more versions of this product

Catalog No. NB122137


Add to cart

Description

Description

MRP1 Monoclonal specifically detects MRP1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

MRP1
MRP1/1343
Unconjugated
10 mM PBS with 0.05% BSA
ABC29, ABCC, ATP-binding cassette sub-family C member 1, ATP-binding cassette, sub-family C (CFTR/MRP), member 1, EC 3.6.3, EC 3.6.3.44, GS-X, Leukotriene C(4) transporter, LTC4 transporter, MRP1DKFZp781G125, MRPDKFZp686N04233, multidrug resistance associated protein 1, multidrug resistance protein, multidrug resistance-associated protein 1, multiple drug resistance protein 1, multiple drug resistance-associated protein
Recombinant full-length human MRP1 protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Western Blot, Flow Cytometry, ELISA
0.2 mg/ml
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA
Purified
Mouse
IgG1 Kappa
100 μg
ABC Transporters, Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
Primary
4363
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit