Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRP1 Mouse anti-Human, Clone: MRP1/1343, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen MRP1
Clone MRP1/1343
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA
Applications Western Blot, Flow Cytometry, ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122137
View Documents
Novus Biologicals
NBP30733210UG
100 μg
Each for $352.89
Add to cart
 
NB122138
View Documents
Novus Biologicals
NBP30733220UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

MRP1 Monoclonal antibody specifically detects MRP1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications

Specifications

MRP1
0.2 mg/ml
Western Blot, Flow Cytometry, ELISA
Purified
RUO
Human
ABC29, ABCC, ATP-binding cassette sub-family C member 1, ATP-binding cassette, sub-family C (CFTR/MRP), member 1, EC 3.6.3, EC 3.6.3.44, GS-X, Leukotriene C(4) transporter, LTC4 transporter, MRP1DKFZp781G125, MRPDKFZp686N04233, multidrug resistance associated protein 1, multidrug resistance protein, multidrug resistance-associated protein 1, multiple drug resistance protein 1, multiple drug resistance-associated protein
Recombinant full-length human MRP1 protein
Monoclonal
Protein A or G purified
MRP1/1343
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA
Unconjugated
Mouse
ABC Transporters, Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
10 mM PBS with 0.05% BSA
4363
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit