Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRP1 Mouse anti-Human, Clone: MRPm6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP1423400.5ML

 View more versions of this product

Catalog No. NBP14234005


Add to cart

Description

Description

MRP1 Monoclonal antibody specifically detects MRP1 in Human samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications

Specifications

MRP1
MRPm6
Western Blot 1:20-1:50, Flow Cytometry 1:20-1:50, Immunohistochemistry 1:20-50, Immunohistochemistry-Paraffin 1:20-50, Immunohistochemistry-Frozen 1:20-50
Supernatant
ABCC1
The antibody reacts with the MRP1 antigen.
0.5 mL
Primary
4363
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Tissue culture supernatant with 0.1% sodium azide
ABC29, ABCC, ATP-binding cassette sub-family C member 1, ATP-binding cassette, sub-family C (CFTR/MRP), member 1, EC 3.6.3, EC 3.6.3.44, GS-X, Leukotriene C(4) transporter, LTC4 transporter, MRP1DKFZp781G125, MRPDKFZp686N04233, multidrug resistance associated protein 1, multidrug resistance protein, multidrug resistance-associated protein 1, multiple drug resistance protein 1, multiple drug resistance-associated protein
Mouse
IgG1
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
MRPm6 reacts with an internal epitope of MRP, a 180-195 kD transmembrane transporter protein overexpressed in various human non-P-glycoprotein MDR tumor cell lines. MRPm6 was raised against a bacterial fusion protein of MRP, containing a segment of 170 amino acids in the carboxy terminal end and part of the carboxy proximal nucleotide binding domain of the protein. MRPm6 does not cross react with the human MDR1 and MDR3 gene products (Flens et al. 1994)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit