Learn More
MRP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 248931AP
Catalog No. 50-173-4600
Add to cart
Description
Description
This gene encodes the prototype of the membrane-associated guanylate kinase (MAGUK) family proteins. MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intercellular junctions. The encoded protein is an extensively palmitoylated membrane phosphoprotein containing a PDZ domain, a Src homology 3 (SH3) motif, and a guanylate kinase domain. This gene product interacts with various cytoskeletal proteins and cell junctional proteins in different tissue and cell types, and may be involved in the regulation of cell shape, hair cell development, neural patterning of the retina, and apico-basal polarity and tumor suppression pathways in non-erythroid cells. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
MRP2
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8VI47, Q92887
ABC30, ABCC2, MRP2
Rabbit
IgG
150 μL
-20°C
Primary
1244, 12780
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ABCC2
Liquid
ABCC2
MRP2 Fusion Protein Ag18694
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse