Learn More
MRPL11 Rabbit anti-Human, Non-human primate, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA551006
Catalog No. PA551006
Add to cart
Description
Description
MRPL11 Polyclonal Antibody detects endogenous levels of MRPL11 protein.
This nuclear gene encodes a 39S subunit component of the mitochondial ribosome. Alternative splicing results in multiple transcript variants. Pseudogenes for this gene are found on chromosomes 5 and 12. [provided by RefSeq, May 2014]
Specifications
Specifications
MRPL11
Polyclonal
Unconjugated
MRPL11
Liquid
MRPL11
Synthesized peptide derived from the Internal region of human MRP-L11.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Non-human Primate
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9Y3B7
MRPL11, CGI-113, 39S ribosomal protein L11, mitochondrial, L11mt, MRP-L11
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
65003