Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL14 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549938

Catalog No. PA549938


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total MRPL14 protein.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found at 17p13.3.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MRPL14
Polyclonal
Unconjugated
Mrpl14
1110006I11Rik; 39S ribosomal protein L14, mitochondrial; 39S ribosomal protein L32, mitochondrial; AI846816; L14mt; L32mt; Mitochondrial large ribosomal subunit protein uL14m; mitochondrial ribosomal protein L14; mitochondrial ribosomal protein L32; Mrpl14; MRP-L14; MRPL32; MRP-L32; RMPL32; Rpml32
Rabbit
Antigen affinity chromatography
RUO
301250, 64928, 68463
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6P1L8, Q7M0E7, Q9D1I6
Mrpl14
Synthesized peptide derived from internal of human MRPL14.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.