Learn More
MRPL14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150401AP
Catalog No. 50-172-9924
Add to cart
Description
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found at 17p13.3.
Specifications
Specifications
MRPL14
0.24 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6P1L8, Q7M0E7, Q9D1I6
L14mt, L32mt, MRP L14, MRP L32, MRPL14, MRPL32, RMPL32, RPML32
Rabbit
IgG
150 μL
-20°C
Primary
301250, 64928, 68463
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MRPL14
Liquid
MRPL14
MRPL14 Fusion Protein Ag6808
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat