Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL16 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549940
Description
The antibody detects endogenous levels of total MRPL16 protein.
MRPL16 Polyclonal antibody specifically detects MRPL16 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
MRPL16 | |
Polyclonal | |
Unconjugated | |
MRPL16 | |
2310039D06Rik; 39S ribosomal protein L16, mitochondrial; L16mt; mitochondiral ribosomal protein L16; Mitochondrial large ribosomal subunit protein uL16m; mitochondrial ribosomal protein L16; Mrpl16; MRP-L16; PNAS-111; rm16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54948 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NX20 | |
MRPL16 | |
Synthesized peptide derived from internal of human MRPL16. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction