Learn More
MRPL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 164921AP
Catalog No. 50-173-0985
Add to cart
Description
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the EcoL2 ribosomal protein family. A pseudogene corresponding to this gene is found on chromosome 12q. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
Specifications
MRPL2
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q498T4, Q5T653, Q9D773
CGI 22, L2mt, MRP L14, MRP L2, MRPL2, RPML14
Rabbit
IgG
150 μL
-20°C
Primary
27398, 301240, 51069
Western Blot
Unconjugated
MRPL2
Liquid
MRPL2
MRPL2 Fusion Protein Ag9618
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat