Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL22 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109977
Description
Immunogen sequence: LAQLEFNDKK GAKIIKEVLL EAQDMAVRDH NVEFRSNLYI AESTSGRGQC LKRIRYHGRG RFGIMEKVYC HYFVKLVEGP PPPPEPPKTA VAHAKEYIQQ LRSRTIVHTL.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the L22 ribosomal protein family. A pseudogene corresponding to this gene is found on chromosome 4q. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
MRPL22 | |
Polyclonal | |
Unconjugated | |
MRPL22 | |
39S ribosomal protein L22, mitochondrial; 39S ribosomal protein L25, mitochondrial; E030011D16Rik; HSPC158; L22mt; L25mt; Mitochondrial large ribosomal subunit protein uL22m; mitochondrial ribosomal protein L22; Mrpl22; MRP-L22; MRPL25; MRP-L25; RPML25 | |
Rabbit | |
Affinity chromatography | |
RUO | |
216767, 287302 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.78 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P0C2C0, Q8BU88 | |
MRPL22 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 97-206 of human MRPL22 (NP_054899.2). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction