Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL35 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101361
Description
Antibody detects endogenous levels of total MRPL35.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Sequence analysis identified three transcript variants. Pseudogenes corresponding to this gene are found on chromosomes 6p, 10q, and Xp. [provided by RefSeq, Jul 2008].
Specifications
MRPL35 | |
Polyclonal | |
Unconjugated | |
MRPL35 | |
1110066C01Rik; 39S ribosomal protein L35, mitochondrial; BM-007; L35mt; Mitochondrial large ribosomal subunit protein bL35m; mitochondrial ribosomal protein L35; MRPL35; MRP-L35; rm35 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51318, 66223 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9CQL6, Q9NZE8 | |
MRPL35 | |
A synthesized peptide derived from human MRPL35(Accession Q9NZE8), corresponding to amino acid residues V107-L157. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction