Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL37 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555265
Description
Immunogen sequence: SATWNRESLL LQVRGSGGAR LSTKDPLPTI ASREEIEATK NHVLETFYPI SPIIDLHECN IYDVK Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit.
Specifications
MRPL37 | |
Polyclonal | |
Unconjugated | |
MRPL37 | |
2300004O14Rik; 39S ribosomal protein L2, mitochondrial; 39S ribosomal protein L37, mitochondrial; AI132596; HSPC235; L2mt; L37mt; MGC878; Mitochondrial large ribosomal subunit protein mL37; mitochondrial ribosomal protein L37; MRPL2; MRP-L2; MRPL37; MRP-L37; ribosomal protein, mitochondrial, L2; RPML2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51253 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZE1 | |
MRPL37 | |
Recombinant protein corresponding to Human MRPL37. Recombinant protein control fragment (Product #RP-94069). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction