Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPL42 Mouse anti-Human, PerCP, Clone: 3H6H2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237385PCP

Catalog No. NBP237385W

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

MRPL42
3H6H2
Flow Cytometry
Purified
MRPL42
Purified recombinant fragment of human MRPL42 (AA: 10-142) expressed in E. Coli.
Protein A or G purified
Stem Cell Markers
Primary
28977
Flow Cytometry
PerCP
PBS with 0.05% Sodium Azide
39S ribosomal protein L31, mitochondrial, 39S ribosomal protein L42, mitochondrial, HSPC204,28S ribosomal protein S32, mitochondrial, L31mt, mitochondrial ribosomal protein L42, mitochondrial ribosomal protein S32, MRPL31L42mt, MRP-L31MRP-S32, MRPS32MRP-L42, PTD007, RPML31S32mt
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit