Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPL42 Mouse anti-Human, Clone: OTI1A4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527137

Catalog No. PIMA527137


Add to cart

Description

Description

Mitochondrial ribosomes consist of a large 39S subunit and a small 28S subunit, both of which are comprised of multiple mitochondrial ribosomal proteins (MRPs) that are encoded by nuclear genes and are essential for protein synthesis within mitochondria. MRP-L42 (mitochondrial ribosomal protein L42), also known as MRP-S32 and MRP-L31, is a 142 amino acid protein that localizes to the mitochondrion, where it exists as a component of the 39S and 28S ribosomal subunits and works in conjunction with other MRPs to mediate protein synthesis. The gene encoding MRP-L42 maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noo syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders.
Specifications

Specifications

MRPL42
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y6G3
39S ribosomal protein L42, mitochondrial; L42mt; MRP-L42; 28S ribosomal protein S32, mitochondrial; MRP-S32; S32mt; 39S ribosomal protein L31, mitochondrial; L31mt; MRP-L31; MRPL42; MRPL31; MRPS32; RPML31
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
28977
Immunohistochemistry (Paraffin), Western Blot
OTI1A4
Unconjugated
MRPL42
Liquid
MRPL42
Full length human recombit protein of MRPL42 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit