Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL44 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549943
Description
The antibody detects endogenous levels of total MRPL44 protein.
MRPL44 Polyclonal antibody specifically detects MRPL44 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
MRPL44 | |
Polyclonal | |
Unconjugated | |
MRPL44 | |
1810030E18Rik; 39S ribosomal protein L44, mitochondrial; 5730593H20Rik; AI463302; COXPD16; l44; L44mt; Mitochondrial large ribosomal subunit protein mL44; mitochondrial ribosomal protein L44; MRPL44; MRP-L44; RM44 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
301552, 65080, 69163 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9CY73, Q9H9J2 | |
MRPL44 | |
Synthesized peptide derived from internal of human MRPL44. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction