Learn More
MRPL46 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 166111AP
Catalog No. 50-173-1053
Add to cart
Description
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
Specifications
Specifications
MRPL46
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5RK00, Q9EQI8, Q9H2W6
C15orf4, L46mt, LIECG2, MRP L46, MRPL46, P2ECSL
Rabbit
IgG
150 μL
-20°C
Primary
26589, 293054, 67308
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MRPL46
Liquid
Mrpl46
MRPL46 Fusion Protein Ag9837
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat