Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPL9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552581
Description
Immunogen sequence: NAPDLACNFS LSQNRGTVIV ERWWKVPLAG EGRKPRLHRR HRVYKLVEDT KHRPKENLEL ILTQSVENVG VRGDLVSVKK SLGRNRLLPQ GLAVYASPEN KKLFEEEKLL RQEGKLEKIQ TKAGEATVKF LKSCRLEVGM KNNV Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 85%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found at 8q21.11.
Specifications
MRPL9 | |
Polyclonal | |
Unconjugated | |
MRPL9 | |
39S ribosomal L9; 39S ribosomal protein L9, mitochondrial; 8030480E20Rik; AA409733; C330013D18Rik; L9mt; Mitochondrial large ribosomal subunit protein bL9m; mitochondrial ribosomal protein L9; Mrpl9; MRP-L9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
65005 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BYD2 | |
MRPL9 | |
Recombinant protein corresponding to Human MRPL9. Recombinant protein control fragment (Product #RP-89169). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction