Learn More
MRPP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 209591AP
Catalog No. 50-173-3174
Add to cart
Description
Description
KIAA0391, also known as MRPP3 (mitochondrial ribonuclease P protein 3), is a 583 amino acid protein that is located in the mitochondria. KIAA0391 functions in tRNA maturation and is a component of mitochondrial ribonuclease P which cleaves tRNA at the 5 ft.-end. Mitochondrial ribonuclease P is an enzyme composed of RG9MTD1, MRPP22 and KIAA0391 subunits. Due to alternative splicing events, KIAA0391 exists as four isoforms. The gene encoding KIAA0391 maps to human chromosome 14, which contains about 700 genes and makes up about 3.5% of human cellular DNA.
Specifications
Specifications
MRPP3
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
B5DF07, O15091, Q8JZY4
KIAA0391, MRPP3
Rabbit
IgG
150 μL
-20°C
Primary
299050, 66132, 9692
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
KIAA0391
Liquid
Kiaa0391, Mrpp3
MRPP3 Fusion Protein Ag15196
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat