Learn More
MRPS10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 160301AP
Catalog No. 50-173-0659
Add to cart
Description
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S10P family. Pseudogenes corresponding to this gene are found on chromosomes 1q, 3p, and 9p.
Specifications
Specifications
MRPS10
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P82664, Q7TQ82, Q80ZK0
MRP S10, MRPS10, PNAS 122, S10mt
Rabbit
IgG
150 μL
-20°C
Primary
363187, 55173, 64657
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
MRPS10
Liquid
Mrps10
MRPS10 Fusion Protein Ag8897
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat