Learn More
Aviva Systems Biology Corp MRPS15 Antibody - middle region (ARP57897_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP57897_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPS15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MRPS15. Alternate names: DC37, FLJ11564, MPR-S15, RPMS15, S15mtApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62170