Learn More
MRPS17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 188811AP
Catalog No. 50-173-2265
Add to cart
Description
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S17P family. The encoded protein is moderately conserved between human mitochondrial and prokaryotic ribosomal proteins. Pseudogenes corresponding to this gene are found on chromosomes 1p, 3p, 6q, 14p, 18q, and Xq.
Specifications
Specifications
MRPS17
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9CQE3, Q9Y2R5
HSPC011, MRP S17, MRPS17, RPMS17, S17mt
Rabbit
IgG
150 μL
-20°C
Primary
288621, 51373, 66258
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MRPS17
Liquid
Mrps17
MRPS17 Fusion Protein Ag13414
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat