Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MRPS18C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116838
Description
Positive Samples: U-87MG, HepG2, 293T Immunogen sequence: MAAVVAVCGG LGRKKLTHLV TAAVSLTHPG THTVLWRRGC SQQVSSNEDL PISMENPYKE PLKKCILCGK HVDYKNVQLL SQFVSPFTGC IYGRHITGLC GKKQKEITKA IKRAQIMGFM PVTYKDPAYL KDPKVCNIRY RE.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S18P family. The encoded protein is one of three that has significant sequence similarity to bacterial S18 proteins. The primary sequences of the three human mitochondrial S18 proteins are no more closely related to each other than they are to the prokaryotic S18 proteins. Pseudogenes corresponding to this gene are found on chromosomes 8p, 12p, 15q, and 22q. [provided by RefSeq, Jul 2008].
Specifications
MRPS18C | |
Polyclonal | |
Unconjugated | |
MRPS18C | |
1110037D14Rik; 28S ribosomal protein S18-1, mitochondrial; 28S ribosomal protein S18c, mitochondrial; CGI-134; mitochondrial ribosomal protein S18-1; mitochondrial ribosomal protein S18C; Mitochondrial small ribosomal subunit protein bS18c; Mitochondrial small ribosomal subunit protein bS18m; MRP-S18-1; MRPS18-1; MRPS18C; MRP-S18-c; mrps18-c; S18mt-c | |
Rabbit | |
Affinity Chromatography | |
RUO | |
289469, 51023, 68735 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2.07 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8R2L5, Q9Y3D5 | |
MRPS18C | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-142 of human MRPS18C (NP_0571511). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction